Sodium hydroxide solution 0.1 N (0.1 M), in single-use containers, HyClone™ for bioprocessing

Supplier: HyClone products (Cytiva)

Synonyms: Caustic soda

SH30883.01 SH30883.03 SH30883.04
SH30883.01EA 351 EUR
SH30883.01 SH30883.03 SH30883.04
Sodium hydroxide solution 0.1 N (0.1 M), in single-use containers, HyClone™ for bioprocessing
Sodium hydroxide

HyClone™ NaOH solutions in single-use bags offer a convenient format to effectively modify pH and properly care for and clean your chromatography resins.


  • Manufactured in ISO 9001 and ISO 13485 certified facility.
  • Lot-specific certificate of analysis (CoA) for each lot produced.
  • Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
  • 0.1 µm sterile filtered.
  • All unit sizes are filled in single-use, ready-to-use bags.
  • Raw materials are USP/NF and EP compendial grade.


HyClone™ NaOH in single-use bags is well-suited for use with: Cytiva's chromatography resins.
Available in 0.1 M (0.1 N), 0.5 M (0.5 N), and 1.0 M (1.0 N) sodium hydroxide concentrations up to 200 L.

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
Merck Index: 14,08627

Order Now

Specification Test Results

Storage 15° to 30°C
Certifications/Compliance Manufactured in a ISO9001 and ISO13485 certified facility
Shelf life 12 Months
pH For Information Only
For use with Chromatography resins, IC systems, ÄKTA hardware systems
Appearance Clear, colorless solution
Sterility No Growth


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR