EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride), Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: N'-(Ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamine monohydrochloride, N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide monohydrochloride, 1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride, EDC hydrochloride, EDC, EDAC HCl, EDAC hydrochloride, 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, WSC HCl, EDC HCl

M-Clarity™ quality level = MQ200

03449-1G 03449-25G 03449-5G
SIAL03449-1GEA 77.1 EUR
SIAL03449-1G SIAL03449-25G SIAL03449-5G
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride), Sigma-Aldrich®
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride
Formula: C₈H₁₇N₃·HCl
Melting Pt: 112…113 °C
Storage Temperature: Freezer
MDL Number: MFCD00012503
CAS Number: 25952-53-8
EINECS: 247-361-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25d8bf2864bcf21ffe6dcca6f22388d1fe427e22&vid=9080dd6b66a1622ef32a2da2138f85e44cc881b8&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR