Agar, Millipore®

Supplier: Merck

Synonyms: Agar Agar

M-Clarity™ quality level = MQ100

05038-500G
SIAL05038-500GEA 289 EUR
SIAL05038-500G
Agar, Millipore®
Agar
Formula: C₁₂nH₁₈nOn₉n
Melting Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS Number: 9002-18-0
EINECS: 232-658-1
Merck Index: 13,00184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR