Diammonium hydrogen citrate, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Citric acid diammonium salt, Diammonium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate, Ammonium citrate dibasic

M-Clarity™ quality level = MQ200

09833-100G 09833-500G
SIAL09833-100GEA 83.5 EUR
SIAL09833-100G SIAL09833-500G
Diammonium hydrogen citrate, Sigma-Aldrich®
di-Ammonium hydrogen citrate
Formula: HOC(CO₂H)(CH₂CO₂NH₄)₂
Density: 1.483 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013068
CAS Number: 3012-65-5
EINECS: 221-146-3
Merck Index: 13,00513

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR