α,α'-Dichloro-p-xylene, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1,4-Bis(chloromethyl)benzene, p-Xylene dichloride, p-Xylylene dichloride

M-Clarity™ quality level = MQ100

105740-100G 105740-25G
SIAL105740-100GEA 101 EUR
SIAL105740-100G SIAL105740-25G
α,α'-Dichloro-p-xylene, Sigma-Aldrich®
α,α'-Dichloro-p-xylene
Formula: C₈H₈Cl₂
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 100 °C
Density: 1.432 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000920
CAS Number: 623-25-6
EINECS: 210-782-7
UN: 2928
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR