1,2-Bis(diphenylphosphino)ethane, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

106496-10G 106496-50G
SIAL106496-10GEA 58.4 EUR
SIAL106496-10G SIAL106496-50G
1,2-Bis(diphenylphosphino)ethane, Sigma-Aldrich®
1,2-Bis(diphenylphosphino)ethane
Formula: C₂₆H₂₄P₂
Melting Pt: 140…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00003047
CAS Number: 1663-45-2
EINECS: 216-769-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR