α,4-Dichlorotoluene, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-Chloro-4-(chloromethyl)benzene, 4-Chlorobenzyl chloride

111961-100G 111961-500G
SIAL111961-100GEA 82 EUR
SIAL111961-100G SIAL111961-500G
α,4-Dichlorotoluene, Sigma-Aldrich®
α,4-Dichlorotoluene
Formula: C₇H₆Cl₂
Boiling Pt: 214 °C (1013 hPa)
Melting Pt: 28…30 °C
Density: 1.259 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000914
CAS Number: 104-83-6
EINECS: 203-242-7
UN: 3427
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR