ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]), Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: ABTS, Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulphonate), 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulphonic acid) diammonium salt

M-Clarity™ quality level = MQ200

11557-1G
SIAL11557-1GEA 108 EUR
SIAL11557-1G
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]), Sigma-Aldrich®
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])
Formula: C₁₈H₂₄N₆O₆S₄
MW: 548.68 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00010404
CAS Number: 30931-67-0
EINECS: 250-396-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR