1,2-Propylenediamine, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 1,2-Diaminopropane

M-Clarity™ quality level = MQ200

117498-100G 117498-500G 117498-2KG
SIAL117498-100GEA 24.7 EUR
SIAL117498-100G SIAL117498-500G SIAL117498-2KG
1,2-Propylenediamine, Sigma-Aldrich®
1,2-Propylenediamine
Formula: C₃H₁₀N₂
Boiling Pt: 120.5 °C (1013 hPa)
Melting Pt: –37 °C
Density: 0.8584 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008089
CAS Number: 78-90-0
EINECS: 201-155-9
UN: 2258
ADR: 8,II
Merck Index: 12,08037

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR