Trioctylphosphine, Sigma-Aldrich®

Supplier: Merck

Synonyms: TOP, Tri-n-octylphosphine

117854-100ML 117854-4X25ML 117854-500ML
SIAL117854-100MLEA 156 EUR
SIAL117854-100ML SIAL117854-4X25ML SIAL117854-500ML
Trioctylphosphine, Sigma-Aldrich®
Trioctylphosphine
Formula: C₂₄H₅₁P
Boiling Pt: 150…155 °C (0.03 mmHg)
Melting Pt: 48 °C
Density: 0.831 g/cm³ (20 °C)
MDL Number: MFCD00015298
CAS Number: 4731-53-7
EINECS: 225-234-2
UN: 1760
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR