4-Biphenylylmethanol, Sigma-Aldrich®

Supplier: Merck

Synonyms: Biphenyl-4-methanol, [1,1'-Biphenyl]-4-ylmethanol, 4-Phenylbenzyl alcohol, 4-Biphenylmethanol

M-Clarity™ quality level = MQ100

123838-10G
SIAL123838-10GEA 49.8 EUR
SIAL123838-10G
4-Biphenylylmethanol, Sigma-Aldrich®
4-Biphenylyl-methanol
Formula: C₁₃H₁₂O
Boiling Pt: 183…184 °C (11 mmHg)
Melting Pt: 104…105 °C
MDL Number: MFCD00004660
CAS Number: 3597-91-9
EINECS: 222-745-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR