1-Methylpiperazine, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

130001-100G 130001-500G 130001-5G
SIAL130001-100GEA 40.5 EUR
SIAL130001-100G SIAL130001-500G SIAL130001-5G
1-Methylpiperazine, Sigma-Aldrich®
1-Methylpiperazine
Formula: C₅H₁₂N₂
Boiling Pt: 138 °C (1013 hPa)
Melting Pt: –6 °C
Density: 0.9085 g/cm³ (20 °C)
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00005966
CAS Number: 109-01-3
EINECS: 203-639-5
UN: 2734
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR