Pyridine-N-oxide, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

131652-100G 131652-25G
SIAL131652-100GEA 81.7 EUR
SIAL131652-100G SIAL131652-25G
Pyridine-N-oxide, Sigma-Aldrich®
Pyridine N-oxide
Formula: C₅H₅NO
Boiling Pt: 270 °C (1013 hPa)
Melting Pt: 61…64 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006194
CAS Number: 694-59-7
EINECS: 211-774-6
Merck Index: 12,08155

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR