3-Hydroxy-p-anisaldehyde, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

143685-100G 143685-25G
SIAL143685-100GEA 93.4 EUR
SIAL143685-100G SIAL143685-25G
3-Hydroxy-p-anisaldehyde, Sigma-Aldrich®
3-Hydroxy-p-anisaldehyde
Formula: C₈H₈O₃
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 68…70 °C
Density: 1.401 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003369
CAS Number: 621-59-0
EINECS: 210-694-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR