2-Methoxythiophenol, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

184055-10G 184055-2G
SIAL184055-10GEA 166 EUR
SIAL184055-10G SIAL184055-2G
2-Methoxythiophenol, Sigma-Aldrich®
2-Methoxythiophenol
Formula: C₇H₈OS
Boiling Pt: 225 °C (1013 hPa)
Density: 1.14 g/cm³ (30 °C)
MDL Number: MFCD00004834
CAS Number: 7217-59-6
EINECS: 230-605-7
UN: 3334
ADR: 9,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR