2-Bromobenzyl alcohol, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

184276-10G 184276-50G
SIAL184276-10GEA 58.8 EUR
SIAL184276-10G SIAL184276-50G
2-Bromobenzyl alcohol, Sigma-Aldrich®
2-Bromobenzyl alcohol
Formula: C₇H₇BrO
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 80…82 °C
MDL Number: MFCD00004600
CAS Number: 18982-54-2
EINECS: 242-719-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR