α,α,α-Trifluoro-m-toluic acid, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 3-(Trifluoromethyl)benzoic acid

M-Clarity™ quality level = MQ100

188344-10G
SIAL188344-10GEA 68.8 EUR
SIAL188344-10G
α,α,α-Trifluoro-m-toluic acid, Sigma-Aldrich®
α,α,α-Trifluoro-m-toluic acid
Formula: C₈H₅F₃O₂
Boiling Pt: 238 °C (1013 hPa)
Melting Pt: 106 °C
Density: 1.371 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002519
CAS Number: 454-92-2
EINECS: 207-230-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR