3-Aminobenzyl alcohol, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-(Hydroxymethyl)aniline, 3-(Hydroxymethyl)aniline, 3-Aminobenzyl Alcohol, (3-Aminophenyl)methanol

M-Clarity™ quality level = MQ100

191396-10G 191396-25G
SIAL191396-10GEA 111 EUR
SIAL191396-10G SIAL191396-25G
3-Aminobenzyl alcohol, Sigma-Aldrich®
3-Aminobenzyl alcohol
Formula: C₇H₉NO
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 90…93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007817
CAS Number: 1877-77-6
EINECS: 217-514-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR