1-Bromo-4-(2-chloroethoxy)benzene, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-Bromo-β-chlorophenetol, 4-Bromo-ω-chlorophenetol, 4-Bromophenyl 2-chloroethyl ether

M-Clarity™ quality level = MQ100

198757-25G
SIAL198757-25GEA 438 EUR
SIAL198757-25G
1-Bromo-4-(2-chloroethoxy)benzene, Sigma-Aldrich®
1-Bromo-4-(2-chloroethoxy)benzene
Formula: C₈H₈BrClO
MDL Number: MFCD00000969
CAS Number: 55162-34-0
EINECS: 259-507-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR