Selenium dioxide, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

200107-25G 200107-5G
SIAL200107-25GEA 154 EUR
SIAL200107-25G SIAL200107-5G
Selenium dioxide, Sigma-Aldrich®
Selenium dioxide
Formula: SeO₂
Melting Pt: 315 °C
Density: 3.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003562
CAS Number: 7446-08-4
EINECS: 231-194-7
UN: 3283
ADR: 6.1,II
Merck Index: 13,08508

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR