Nickel sulphate heptahydrate, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

203890-10G 203890-50G
SIAL203890-10GEA 74.8 EUR
SIAL203890-10G SIAL203890-50G
Nickel sulphate heptahydrate, Sigma-Aldrich®
Nickel sulphate heptahydrate
Formula: NiO₄S·7H₂O
Melting Pt: 99 °C
Density: ∼2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149814
CAS Number: 10101-98-1
EINECS: 232-104-9
UN: 3077
ADR: 9,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR