Ytterbium(III) chloride hexahydrate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

204870-10G 204870-50G
SIAL204870-10GEA 242 EUR
SIAL204870-10G SIAL204870-50G
Ytterbium(III) chloride hexahydrate, Sigma-Aldrich®
Ytterbium (III) chloride hexahydrate
Formula: Cl₃Yb·6H₂O
Melting Pt: 154 °C
Density: 2.57 g/cm³ (20 °C)
MDL Number: MFCD00149885
CAS Number: 10035-01-5
EINECS: 233-800-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR