Zirconium(IV) oxide, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Zirconium dioxide, Zirconium Oxide

M-Clarity™ quality level = MQ100

204994-10G
SIAL204994-10GEA 151 EUR
SIAL204994-10G
Zirconium(IV) oxide, Sigma-Aldrich®
Zirconium (IV) oxide
Formula: ZrO₂
MW: 123.22 g/mol
Boiling Pt: 4300
Melting Pt: 2680
Density: 5.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011310
CAS Number: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR