Praseodymium(III) nitrate hexahydrate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Praseodymium trinitrate hexahydrate

M-Clarity™ quality level = MQ200

205133-10G 205133-250G 205133-50G
SIAL205133-10GEA 60.6 EUR
SIAL205133-10G SIAL205133-250G SIAL205133-50G
Praseodymium(III) nitrate hexahydrate, Sigma-Aldrich®
Praseodymium trinitrate hexahydrate
Formula: N₃O₉Pr·6H₂O
Melting Pt: 56 °C
Density: 2.233 g/cm³ (20 °C)
MDL Number: MFCD00149826
CAS Number: 15878-77-0
EINECS: 233-796-5
UN: 1477
ADR: 5.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR