Nitric acid 70% 70, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Nitric acid solution
MQ100
225711-475ML
SIAL225711-475MLEA 150 EUR
SIAL225711-475ML
Nitric acid 70% 70, Sigma-Aldrich®
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 87
Melting Pt: -42
Density: 1.51
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 I
Merck Index: 13,06610