di-Sodium tetraborate, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

229946-100G 229946-25G 229946-5G
SIAL229946-100GEA 232 EUR
SIAL229946-100G SIAL229946-25G SIAL229946-5G
di-Sodium tetraborate, Sigma-Aldrich®
di-Sodium tetraborate
Formula: Na₂B₄O₇
Boiling Pt: 1575 °C (1013 hPa)
Melting Pt: 742 °C
Density: 2.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081185
CAS Number: 1330-43-4
EINECS: 215-540-4
REACH: 01-2119490790-32
Merck Index: 12,08733

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR