2,4,6-Trichloroanisole, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 1,3,5-Trichloro-2-methoxybenzene, 2,4,6-Trichloro-1-methoxybenzene

M-Clarity™ quality level = MQ100

235393-1G 235393-5G
SIAL235393-1GEA 53.9 EUR
SIAL235393-1G SIAL235393-5G
2,4,6-Trichloroanisole, Sigma-Aldrich®
2,4,6-Trichloroanisole
Formula: C₇H₅Cl₃O
Boiling Pt: 235…238 °C (1013 hPa)
Melting Pt: 60…61 °C
Density: 1.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000588
CAS Number: 87-40-1
EINECS: 201-743-5
Merck Index: 13,09702

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR