Barium chlorate monohydrate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

244554-500G
SIAL244554-500GEA 57.8 EUR
SIAL244554-500G
Barium chlorate monohydrate, Sigma-Aldrich®
Barium chlorate monohydrate
Formula: BaCl₂O₆·1H₂O
MDL Number: MFCD00149147
CAS Number: 10294-38-9
EINECS: 236-760-7
UN: 1445
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR