Calcium wolframate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Calcium tungstate, Calcium tungsten oxide

M-Clarity™ quality level = MQ100

248665-100G 248665-500G
SIAL248665-100GEA 198 EUR
SIAL248665-100G SIAL248665-500G
Calcium wolframate, Sigma-Aldrich®
Calcium wolframate
Formula: CaO₄W
MDL Number: MFCD00010913
CAS Number: 7790-75-2
EINECS: 232-219-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR