(R)-(-)-α-Methoxyphenylacetic acid, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: (R)-(-)-alpha-Methoxyphenylacetic Acid

M-Clarity™ quality level = MQ100

248967-1G
SIAL248967-1GEA 154 EUR
SIAL248967-1G
(R)-(-)-α-Methoxyphenylacetic acid, Sigma-Aldrich®
(R)-(-)-α-Methoxyphenylacetic acid
Formula: C₉H₁₀O₃
Boiling Pt: 164…166 °C (18 mmHg)
Melting Pt: 64…67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004250
CAS Number: 3966-32-3
EINECS: 223-580-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR