Lactic acid 85% ≥85% in aqueous solution, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid

M-Clarity™ quality level = MQ200

252476-1KG 252476-100G 252476-2.5KG
SIAL252476-1KGEA 126 EUR
SIAL252476-1KG SIAL252476-100G SIAL252476-2.5KG
Lactic acid 85% ≥85% in aqueous solution, Sigma-Aldrich®
Lactic acid
Formula: H₃CCH(OH)COOH
Boiling Pt: 122 °C (20 hPa)
Density: 1.11…1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
EINECS: 200-018-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR