2,5-Dihydrofuran, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

253170-100G 253170-25G
SIAL253170-100GEA 236 EUR
SIAL253170-100G SIAL253170-25G
2,5-Dihydrofuran, Sigma-Aldrich®
2,5-Dihydrofuran
Formula: C₄H₆O
MDL Number: MFCD00003219
CAS Number: 1708-29-8
EINECS: 216-957-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR