Magnesium chloride hexahydrate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Magnesium dichloride

M-Clarity™ quality level = MQ100

255777-100G 255777-25G 255777-5G
SIAL255777-100GEA 739 EUR
SIAL255777-100G SIAL255777-25G SIAL255777-5G
Magnesium chloride hexahydrate, Sigma-Aldrich®
Magnesium chloride hexahydrate
Formula: MgCl₂·6H₂O
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 117 °C
Density: 1.57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR