Disodium hydrogen phosphate, Sigma-Aldrich®

Supplier: Merck

Synonyms: sec-Sodium phosphate, di-Sodium hydrogen orthophosphate, di-Sodium hydrogen phosphate anhydrous, Sodium hydrogen phosphate, Disodium phosphate, Sodium phosphate dibasic

M-Clarity™ quality level = MQ100

255793-10G 255793-50G
SIAL255793-10GEA 152 EUR
SIAL255793-10G SIAL255793-50G
Disodium hydrogen phosphate, Sigma-Aldrich®
di-Sodium hydrogen phosphate
Formula: Na₂HPO₄
Melting Pt: 250 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003496
CAS Number: 7558-79-4
EINECS: 231-448-7
REACH: 01-2119489797-11
Merck Index: 12,08805

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR