Hexaammineruthenium trichloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: Hexaammineruthenium(III) chloride, Hexaammineruthenium (III) chloride

M-Clarity™ quality level = MQ100

262005-1G 262005-250MG 262005-5G
SIAL262005-1GEA 143 EUR
SIAL262005-1G SIAL262005-250MG SIAL262005-5G
Hexaammineruthenium trichloride, Sigma-Aldrich®
Hexaammineruthenium trichloride
Formula: H₁₈Cl₃N₆Ru
MDL Number: MFCD00011478
CAS Number: 14282-91-8
EINECS: 238-176-8
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR