Indium, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

264067-2.8G
SIAL264067-2.8GEA 133 EUR
SIAL264067-2.8G
Indium, Sigma-Aldrich®
Indium
Formula: In
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 156 °C
Density: 7.3 g/cm³ (20 °C)
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR