Arsenic, powder, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Arsenic solution

M-Clarity™ quality level = MQ100

267961-10G
SIAL267961-10GEA 267 EUR
SIAL267961-10G
Arsenic, powder, Sigma-Aldrich®
Arsenic
Formula: As
MW: 74.92 g/mol
Melting Pt: 817
Density: 5.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085309
CAS Number: 7440-38-2
EINECS: 231-148-6
UN: 1558
ADR: 6.1 II
Merck Index: 12,00832

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR