Ethyl oleate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Elaidic acid ethyl ester, (Z)-9-Octadecenoic acid ethyl ester, Oleic acid ethyl ester

M-Clarity™ quality level = MQ100

268011-5G 268011-25G
SIAL268011-5GEA 55.6 EUR
SIAL268011-5G SIAL268011-25G
Ethyl oleate, Sigma-Aldrich®
Ethyl oleate
Formula: C₂₀H₃₈O₂
Boiling Pt: 386 °C (1013 hPa)
Melting Pt: –32 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 174 °C
Storage Temperature: Ambient
MDL Number: MFCD00009579
CAS Number: 111-62-6
EINECS: 203-889-5
Merck Index: 12,06965

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR