Tritolyl phosphate (mixture of isomers), Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Tricresyl phosphate, Tritolyl phosphate, Tris(methylphenyl) phosphate, TCP

M-Clarity™ quality level = MQ100

268917-500ML
SIAL268917-500MLEA 72.9 EUR
SIAL268917-500ML
Tritolyl phosphate (mixture of isomers), Sigma-Aldrich®
Tritolyl phosphate (mixture of isomeres)
Formula: C₂₁H₂₁O₄P
Boiling Pt: 420 °C (1013 hPa)
Melting Pt: –40 °C
Density: 1.16 g/cm³ (20 °C)
MDL Number: MFCD02102116
CAS Number: 1330-78-5
EINECS: 215-548-8
UN: 2810
ADR: 6.1,II
Merck Index: 12,09892

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR