Chromotrope 2B, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

27130-10G
SIAL27130-10GEA 48.5 EUR
SIAL27130-10G
Chromotrope 2B, Sigma-Aldrich®
Chromotrope 2B
Formula: C₁₆H₉N₃Na₂O₁₀S₂
MDL Number: MFCD00003951
CAS Number: 548-80-1
EINECS: 208-959-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR