Perchloric acid ≥70%

Supplier: Merck
Danger

311421-250ML 311421-50ML
SIAL311421-250MLEA 205 EUR
SIAL311421-250ML SIAL311421-50ML
Perchloric acid ≥70%
Perchloric acid
Formula: HClO₄
Boiling Pt: 160…181 °C (1013 hPa)
Melting Pt: –112.2 °C
Density: 1.53…1.768 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 1873
ADR: 5.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 70%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR