5-Methylpyrazine-2-carboxylic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

347647-25G 347647-5G
SIAL347647-25GEA 154 EUR
SIAL347647-25G SIAL347647-5G
5-Methylpyrazine-2-carboxylic acid, Sigma-Aldrich®
5-Methylpyrazine-2-carboxylic acid
Formula: C₆H₆N₂O₂
MDL Number: MFCD00068241
CAS Number: 5521-55-1
EINECS: 413-260-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR