Gold, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

349305-1.5G
SIAL349305-1.5GEA 547 EUR
SIAL349305-1.5G
Gold, Sigma-Aldrich®
Gold
Formula: Au
Boiling Pt: 2800 °C (1013 hPa)
Melting Pt: 1063 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR