Dimethylamine 40% in aqueous solution, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

426458-100ML 426458-1L 426458-2L
SIAL426458-100MLEA 25.6 EUR
SIAL426458-100ML SIAL426458-1L SIAL426458-2L
Dimethylamine 40% in aqueous solution, Sigma-Aldrich®
Dimethylamine
Formula: C₂H₇N
Storage Temperature: Ambient
MDL Number: MFCD00008288
CAS Number: 124-40-3
EINECS: 204-697-4
UN: 1160
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR