(3-Glycidoxypropyl)trimethoxysilane, Sigma-Aldrich®

Supplier: Merck

Synonyms: (3-Glycidyloxypropyl)trimethoxysilane, 3-Glycidoxypropyltrimethoxysilane, (3-(2,3-Epoxypropoxy)propyl)trimethoxysilane

M-Clarity™ quality level = MQ200

440167-100ML 440167-1L 440167-2.5L 440167-500ML
SIAL440167-100MLEA 68.7 EUR
SIAL440167-100ML SIAL440167-1L SIAL440167-2.5L SIAL440167-500ML
(3-Glycidoxypropyl)trimethoxysilane, Sigma-Aldrich®
(3-Glycidoxypropyl)trimethoxysilane
Formula: C₉H₂₀O₅Si
Boiling Pt: 299 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00005144
CAS Number: 2530-83-8
EINECS: 219-784-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR