Dodecyltriethoxysilane, Sigma-Aldrich®

Supplier: Merck

Synonyms: Triethoxy(dodecyl)silane

44237-25ML
SIAL44237-25MLEA 246 EUR
SIAL44237-25ML
Dodecyltriethoxysilane, Sigma-Aldrich®
Dodecyltriethoxysilane
Formula: C₁₈H₄₀O₃Si
MDL Number: MFCD00048437
CAS Number: 18536-91-9
EINECS: 242-409-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR