Graphite, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

496553-180.7G
SIAL496553-180.7GEA 800 EUR
SIAL496553-180.7G
Graphite, Sigma-Aldrich®
Graphite
Formula: C
Melting Pt: 3827 °C
Density: 2.09…2.23 g/cm³ (20 °C)
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR