Triacetin (glycerol triacetate), Sigma-Aldrich®

Supplier: Merck

Synonyms: Glyceryl triacetate, Propane-1,2,3-triyl triacetate, Glycerol triacetate

M-Clarity™ quality level = MQ100

525073-1L 525073-20L 525073-4L
SIAL525073-1LEA 83.1 EUR
SIAL525073-1L SIAL525073-20L SIAL525073-4L
Triacetin (glycerol triacetate), Sigma-Aldrich®
Triacetin (Glycerol triacetate)
Formula: C₉H₁₄O₆
Boiling Pt: 258 °C (1013 hPa)
Melting Pt: 4 °C
Density: 1.1596 g/cm³ (20 °C)
Flash Pt: 138 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008716
CAS Number: 102-76-1
EINECS: 203-051-9
Merck Index: 13,09664

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR