Propagyl bromide ∼9.2 mol/l in xylene ≥80%, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

530409-125G 530409-50G
SIAL530409-125GEA 218 EUR
SIAL530409-125G SIAL530409-50G
Propagyl bromide ∼9.2 mol/l in xylene ≥80%, Sigma-Aldrich®
Propagyl bromide
Formula: C₃H₃Br
MDL Number: MFCD00000241
CAS Number: 106-96-7
EINECS: 203-447-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR