Zirconium(IV) tert-butanolate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Zirconium(IV) t-butoxide, Zirconium (IV) tert-butoxide, Zirconium (IV) tert-butylate

M-Clarity™ quality level = MQ100

560030-25G 560030-5G
SIAL560030-25GEA 1040 EUR
SIAL560030-25G SIAL560030-5G
Zirconium(IV) tert-butanolate, Sigma-Aldrich®
Zirconium (IV) tert-butanolate
Formula: C₁₆H₃₆O₄Zr
MDL Number: MFCD00075085
CAS Number: 2081-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR